Zinc chloride 97.0-100.5%, granular USP, J.T.Baker®
Catalog #: CAJT4320-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7646-85-7
MOC: Regulated LimitedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:PE pail, screw top
- Size:12 kg
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Store at 2 to 25 °C:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- Identification:Passes Test
- Oxychloride:Passes Test
- Sulfate (SO₄):≤0.03 %
- Alkalies and Alkaline Earths:≤1.0 %
- Ammonium Salts:Passes Test
- Lead (Pb):≤0.005 %
- Assay (ZnCl₂):97.0 - 100.5 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
Specification Test Results
About this item
Synonyms: Zinc dichloride,Dichlorozinc
Boiling Pt:732 °C (1013 hPa)
Melting Pt:283 °C
Density:2.91 g/cm³ (20 °C)
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,0-100,5
:Management of Change (MOC) category = RL
:Boiling Pt:
:Boiling Pt: 732 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 283 °C
:Flash Pt:
:Flash Pt:
:M.W. 136.3 g/mol
Merck Index:12,10261
:Zinc chloride
Storage Temperature:Ambient
EINECS:231-592-0
UN:2331
:8
:III
:GHS05
:GHS07
:GHS09
:GHS08
:M.O.C. Category RL
Formula:ZnCl₂
MW:136.3 g/mol
MDL Number:MFCD00011295
CAS Number:7646-85-7
Customers Who Bought This Also Bought