Zinc acetate dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Acetic acid zinc salt dihydrate,Zinc (II) acetate dihydrate
Formula:(H₃CCOO)₂Zn·2H₂O
MW:219.51 g/mol
Melting Pt:237 °C
Density:1.74 g/cm³
MDL Number:MFCD00066961
CAS Number:5970-45-6
ADR:9,III
Merck Index:12,10256
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 237 °C
:Flash Pt:
:Flash Pt:
:M.W. 219.51 g/mol
EINECS:209-170-2
UN:3077
:9
:III
:GHS07
:GHS09
Storage Temperature:Ambient
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay ((CH₃COO)₂Zn·2H₂O):99.0 - 101.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - pH of 5% Solution at 25°C:6.0 - 7.0
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Lead (Pb):≤0.002 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- J.T.Baker®, Zinc acetate dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number4296 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR4296
SDS
spec_JT4296-1 spec_JT4296-5
Product specification
Customers Who Bought This Also Bought