Specifications
- Description:VWR® Hazardous location undercounter freezer
- Defrost:Manual
- Door type:One swing solid door
- Shelves/drawers:2 Fixed shelves/3 removable baskets
- Weight:41 kg (91 lbs.)
- Exterior W×D×H:600×600×819 mm (23⁵/₈×23⁵/₈×32¹/₄")
- Interior W×D×H:483×406×622 mm (19×16×24¹/₂")
- Capacity:113 L (4 cu.ft.)
- Operating Temperature:–15...–25 °C
Specifications
About this item
Designed for safety with ATEX and non-sparking components, these units are ideal for keeping you and your substances safe.
- Microprocessor temperature control
- ATEX non-sparking components
- Natural hydrocarbon refrigerants
- Meet NFPA and OSHA guidelines for hazardous laboratory locations
In the fast-paced world of scientific and industrial laboratories, precision and safety are key. These hazardous location cold storage units are engineered for use in environments where volatile or explosive conditions could exist. These units meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines. Engineered with microprocessor temperature control, they provide exceptional precision, recovery, and maintenance. You'll also experience the power of natural hydrocarbon refrigerants through energy savings and a greener laboratory footprint - all while enjoying stellar system performance.