Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC, Macron Fine Chemicals™
Catalog #: CAMKV15506
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 64-19-7
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, clear, Mall
- Size:500 ml
- Supplier No.:V155-06
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- Assay (by GC, corrected for water):≥99.7 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤0.4 ppm
- Color (APHA):≤10
- Dilution Test:Passes Test
- Heavy Metals (by ICP-OES):≤5 ppm
- Identification:Passes Test
- Residue after Evaporation:≤5 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Sulfate (SO₄):≤0.4 ppm
- Titrable Base (meq/g):≤0.0004
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.02
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC:
Specification Test Results
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = N
Flash Pt:38.5 °C
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Formula:H₃CCOOH
MW:60.05 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00036152
CAS Number:64-19-7
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:M.O.C. Category N
Customers Who Bought This Also Bought