Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Ammonium hydroxide
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MW:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
MDL Number:MFCD00066650
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 32…37.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –60…–57.5 °C
:Flash Pt:
:Flash Pt:
:M.W. 35.05 g/mol
:Ammonia solution
Storage Temperature:Ambient
EINECS:215-647-6
:32…37.7 °C (1013 hPa)
UN:2672
:8
:III
:GHS05
:GHS07
:GHS09
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (as NH₃):28.0 - 30.0 %
- Appearance:Passes Test
- ACS - Carbon Dioxide (CO₂):≤0.002 %
- ACS - Pyridine:Passes Test
- ACS - Residue after Ignition:≤0.002 %
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Specific Gravity at 60°/60°F:0.896 - 0.902
- Trace Impurities - Arsenic (As):≤3.000 ppm
- Chloride (Cl):≤0.5 ppm
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Total Sulfur (as SO₄):≤2 ppm
- Determine by Flame Photometry & ICP - Aluminum (Al):≤0.100 ppm
- Determine by Flame Photometry & ICP - Barium (Ba):≤0.100 ppm
- Determine by Flame Photometry & ICP - Beryllium (Be):≤0.010 ppm
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤0.010 ppm
- Determine by Flame Photometry & ICP - Boron (B):≤0.050 ppm
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤0.100 ppm
- Determine by Flame Photometry & ICP - Calcium (Ca):≤0.100 ppm
- Determine by Flame Photometry & ICP - Chromium (Cr):≤0.050 ppm
- Determine by Flame Photometry & ICP - Cobalt (Co):≤0.050 ppm
- Determine by Flame Photometry & ICP - Copper (Cu):≤0.050 ppm
- Determine by Flame Photometry & ICP - Gallium (Ga):≤0.500 ppm
- Determine by Flame Photometry & ICP - Germanium (Ge):≤0.500 ppm
- Determine by Flame Photometry & ICP - Gold (Au):≤0.500 ppm
- Determine by Flame Photometry & ICP - Iron (Fe):≤0.100 ppm
- Determine by Flame Photometry & ICP - Lead (Pb):≤0.2 ppm
- Determine by Flame Photometry & ICP - Lithium (Li):≤0.050 ppm
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤0.100 ppm
- Determine by Flame Photometry & ICP - Manganese (Mn):≤0.050 ppm
- Determine by Flame Photometry & ICP - Mercury (Hg):≤0.500 ppm
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤0.010 ppm
- Determine by Flame Photometry & ICP - Nickel (Ni):≤0.500 ppm
- Determine by Flame Photometry & ICP - Potassium (K):≤0.100 ppm
- Determine by Flame Photometry & ICP - Silicon (Si):≤0.100 ppm
- Determine by Flame Photometry & ICP - Silver (Ag):≤0.050 ppm
- Determine by Flame Photometry & ICP - Sodium (Na):≤0.500 ppm
- Determine by Flame Photometry & ICP - Strontium (Sr):≤0.100 ppm
- Determine by Flame Photometry & ICP - Thallium (Tl):≤0.050 ppm
- Determine by Flame Photometry & ICP - Tin (Sn):≤0.100 ppm
- Determine by Flame Photometry & ICP - Vanadium (V):≤0.010 ppm
- Determine by Flame Photometry & ICP - Zinc (Zn):≤0.100 ppm
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤0.010 ppm
- Determine by Flame Photometry & ICP:
- Macron Fine Chemicals™, Ammonia solution 28.0 - 30.0% (NH₃ basis), AR Select® ACS for trace metal analysis:
Specification Test Results
Related Information
Certificates
Search by Lot Number6665 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
sds_MACR6665
SDS
spec_MK666514
Product specification
Customers Who Bought This Also Bought