Ammonia solution 28.0 - 30.0% (NH₃ basis), AR® ACS (26° Baume), Macron Fine Chemicals™
Supplier: Avantor
MOC: Non-Regulated
About this item
Synonyms: Ammonium hydroxide
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MW:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.90 g/cm³
MDL Number:MFCD00066650
ADR:8,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 32…37.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –60…–57.5 °C
:Flash Pt:
:Flash Pt:
:M.W. 35.05 g/mol
:Ammonia solution
Storage Temperature:Ambient
EINECS:215-647-6
:32…37.7 °C (1013 hPa)
UN:2672
:8
:III
:GHS05
:GHS07
:GHS09
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (as NH₃):28.0 - 30.0 %
- Appearance:Passes Test
- ACS - Carbon Dioxide (CO₂):≤0.002 %
- ACS - Residue after Ignition:≤0.002 %
- ACS - Substances Reducing Permanganate:Passes Test
- Arsenic and Antimony (as As):≤3 ppm
- Chloride (Cl):≤0.5 ppm
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤2 ppm
- Trace Impurities - Aluminum (Al):≤0.300 ppm
- Trace Impurities - Boron (B):≤0.100 ppm
- Trace Impurities - Calcium (Ca):≤0.300 ppm
- Trace Impurities - Chromium (Cr):≤0.200 ppm
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Gold (Au):≤0.300 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Lead (Pb):≤0.200 ppm
- Trace Impurities - Magnesium (Mg):≤0.300 ppm
- Trace Impurities - Manganese (Mn):≤0.200 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Trace Impurities - Potassium (K):≤0.300 ppm
- Trace Impurities - Sodium (Na):≤2.000 ppm
- Trace Impurities - Tin (Sn):≤0.300 ppm
- Trace Impurities - Titanium (Ti):≤0.300 ppm
- Trace Impurities - Zinc (Zn):≤0.300 ppm
- Macron Fine Chemicals™, Ammonia solution 28.0 - 30.0% (NH₃ basis), AR® ACS (26° Baume):
Specification Test Results
Related Information
Certificates
Search by Lot Number3256 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Customers Who Bought This Also Bought