Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: 2-Propanone,DMK,Propanone
Formula:CH₃COCH₃
MW:58.08 g/mol
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
MDL Number:MFCD00008765
CAS Number:67-64-1
ADR:3,II
Merck Index:13,00067
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -95.4 °C|Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: <–20 °C (closed cup)
:M.W. 58.08 g/mol
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
REACH:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
:GHS08
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₃COCH₃) (by GC, corrected for water):≥99.5 %
- ACS - Aldehydes (as HCHO):≤0.002 %
- ACS - Color (APHA):≤10
- ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- ACS - Methanol (CH₃OH)(by GC):≤0.05 %
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Solubility in H₂O:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Titrable Base (meq/g):≤0.0006
- ACS - Water (H₂O):≤0.5 %
- Appearance (clear, colorless liquid):
- Date Received: / /:
- Date Opened: / /:
- Macron Fine Chemicals™, Acetone ≥99.5% (by GC, corrected for water content), AR® ACS for histology:
Specification Test Results
Related Information
Certificates
Search by Lot Number2440 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 2440 AVANTOR MACRON 2440
Datasheet
sds_MACR2440
SDS
Customers Who Bought This Also Bought