L(+)-Ascorbic acid 99.0-100.5% USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Ascorbic Acid,(R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one
Formula:C₆H₈O₆
MW:176.13 g/mol
Boiling Pt:553 °C (1013 hPa)
Melting Pt:190…192 °C
Density:1.694 g/cm³ (20 °C)
MDL Number:MFCD00064328
CAS Number:50-81-7
Merck Index:13,00837
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 553 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 190 to 192 °C|Melting Pt: 190…192 °C
:Flash Pt:
:Flash Pt:
:M.W. 176.13 g/mol
:L(+)-Ascorbic acid
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Appearance (fine, white to slightly yellow crystals):Passes Test
- USP - Assay (C₆H₈O₆):99.0 - 100.5 %
- USP - Specific Rotation [Å]²⁵^D (+):20.5 - 21.5 Degree
- USP - Residue on Ignition:≤0.1 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- Residual Solvents - Only Class 2 Solvents (Methanol) and Class 3:
- Solvents are likely to be present. Residual Class 2 Solvents are below:
- Option 1 and the residual concentration of all Class 3 Solvents is:
- <0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number1852 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR1852
SDS
Customers Who Bought This Also Bought