Acetic acid glacial 99.5-100.05% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
New Product
Certificates
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Formula:H₃CCOOH
MW:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
MDL Number:MFCD00036152
CAS Number:64-19-7
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,05
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets B.P. Chemical Specifications:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification:Passes Test
- USP - Congealing Temperature:≥15.6 °C
- USP - Nonvolatile Residue (USP), mg/20 mL:≤1.0
- USP - Chloride (Cl):Passes Test
- USP - Sulfate (SO₄):Passes Test
- USP - Readily Oxidizable Substances:Passes Test
- USP - Assay (CH₃COOH):99.5 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance:Passes Test
- EP/BP - Freezing Point:≥14.8 °C
- EP/BP - Iron (Fe):≤5 ppm
- EP/BP - Chloride (Cl):≤25 ppm
- EP/BP - Sulfate (SO₄):≤50 ppm
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Residue on Evaporation:≤0.01 %
- EP/BP - Assay (CH₃COOH):99.0 - 100.5 %
- JP - Assay (CH₃COOH):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Congealing Point:≥14.5 °C
- JP - Nonvolatile Residue, mg/10 mL:≤1.0
- JP - Chloride (Cl):Passes Test
- JP - Sulfate (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Reducing Substances:Passes Test
- Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or:
- produced by the production process.:
- Preserve in tight containers, and store at room temperature:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Acetic acid glacial 99.5-100.05% USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number9526 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
sds_BAKR9526
SDS
spec_JT9526-1 spec_JT9526-20 spec_JT9526-3 spec_JT9526-7
Product specification
Customers Who Bought This Also Bought