Acetic acid glacial ≥99.7% for HPLC, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Formula:H₃CCOOH
MW:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
MDL Number:MFCD00036152
CAS Number:64-19-7
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- Assay (by GC, corrected for water):≥99.7 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.01
- Residue after Evaporation:≤0.0005 %
- Water (by Karl Fischer titrn):≤0.1 %
- (1) Snyder, L.R., J. Chromatography, 92,223-230 (1974):
- J.T.Baker®, Acetic acid glacial ≥99.7% for HPLC:
Specification Test Results
Related Information
Certificates
Search by Lot Number9515 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor J.T.Baker® LCMS and HPLC Solvents and Reagents Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
sds_BAKR9515
SDS
Customers Who Bought This Also Bought