Acetic acid glacial ≥99.7%, CMOS for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Formula:H₃CCOOH
MW:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
MDL Number:MFCD00036152
CAS Number:64-19-7
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- Assay (CH₃COOH) (by freezing point):≥99.7 %
- Color (APHA):≤10
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Acetaldehyde:≤0.05 %
- Residue after Evaporation:≤4 ppm
- Solubility in H₂O:Passes Test
- Specific Gravity at 20°/20°C:≥1.048
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤1 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate (SO₄):≤0.5 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Arsenic and Antimony (as As):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤50 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤200.0 ppb
- Trace Impurities - Chromium (Cr):≤30.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤20.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Heavy Metals (as Pb):≤300.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤25.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silicon (Si):≤50 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤100.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤50.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- For additional information, go to www.askavantor.com. Search keywords:
- freezing" and product name.":
- J.T.Baker®, Acetic acid glacial ≥99.7%, CMOS for microelectronic:
Specification Test Results
Related Information
Certificates
Search by Lot Number9513 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT9513-7
Product specification
Customers Who Bought This Also Bought