Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Vinegar acid,Methanecarboxylic acid
Formula:H₃CCOOH
MW:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
MDL Number:MFCD00036152
CAS Number:64-19-7
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 118 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 17 °C
:Flash Pt:
:Flash Pt: 38.5 °C
:M.W. 60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
REACH:01-2119475328-30
:8
:II
:GHS02
:GHS05
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:
- ACS - Assay (by GC, corrected for water):≥99.7 %
- ACS - Color (APHA):≤10
- Dilution Test:Passes Test
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- ACS - Chloride (Cl):≤1 ppm
- ACS - Sulfate (SO₄):≤1 ppm
- ACS - Substances Reducing Dichromate:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Base (meq/g):≤0.0004
- ACS - Suitability for Cholesterol Determination:Passes Test
- ACS - Acetaldehyde:≤0.005 %
- ACS - Heavy Metals (by ICP-OES):≤5 ppm
- Trace Impurities - ACS - Iron (Fe):≤0.200 ppm
- J.T.Baker®, Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9511 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Brand Definition Baker Analyzed
Datasheet
Customers Who Bought This Also Bought