Pierce CL-Xposure™ Film, Thermo Scientific
Supplier: Thermo Scientific
CE Compliant
Certificates
About this item
CL-XPosure™ Film is a clear blue X-ray film recommended for the detection and documentation of blotting assays.
- Wide compatibility with various blotting methods
- Ideal for life science research applications
CL-XPosure™ works with chemiluminescent horseradish peroxidase (HRP) or alkaline phosphatase (AP) detection reagents as well as radioactivity, making it ideal for Western, Southern and Northern blots and Electrophoretic Mobility Shift Assays (EMSA). CL-XPosure™ Film exhibits the same detection sensitivity as other commercially available film brands.
Delivery: Supplied in packs of 25, 50 or 100 non-interleaved sheets.
Related Information
Thermo Scientific Reagents for Manual Western Blot Detection Brochure
Product Literature
Customers Who Bought This Also Bought