VWR® Pipet Tip Disposal Box
Catalog #: 82010-754
Supplier: VWR International
Specifications
- Dimensions:15.2×15.2×15.2 cm (6× 6×6") (W×L×H)
- Cat. no.:82010-754
Specifications
About this item
Box features a flip-top port cover for convenient one-handed disposal of used pipet tips.
- Thick acrylic construction
- Protects against beta radiation
- Large, hinged lid
- Easy disposal of used pipet tips
Box features a flip-top port cover for convenient one-handed disposal of used pipet tips.
Eliminates any need to handle pipet tips used with hazardous fluids. The 10mm (3/8") thick acrylic provides protection from beta radiation. Large, hinged lid allows easy removal of contents.
Customers Who Bought This Also Bought