Aluminum etch (80:15:3:2), CMOS, J.T.Baker®
Catalog #: JT5459-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:13 lbs.
- Cat. no.:JT5459-5
- Supplier No.:5459-05
Specifications
Specification Test Results
- For Microelectronic Use:
- Phosphoric Acid (H₃PO₄)(w/w):72.5 - 74.5 %
- Nitric Acid (HNO₃)(w/w):1.7 - 2.3 %
- Acetic Acid (CH₃COOH)(w/w):9.5 - 10.5 %
- Antimony (Sb):≤15.000 ppm
- Calcium (Ca):≤15 ppm
- Chloride (Cl):≤3 ppm
- Sodium (Na):≤10.000 ppm
- Sulfate (SO₄):≤20 ppm
- Insoluble Matter:≤50 ppm
- Arsenic (As):≤0.200 ppm
- Trace Impurities - Copper (Cu):≤1 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤6.000 ppm
- Trace Impurities - Lithium (Li):≤0.300 ppm
- Trace Impurities - Magnesium (Mg):≤2.000 ppm
- Trace Impurities - Manganese (Mn):≤0.200 ppm
- Trace Impurities - Nickel (Ni):≤1.000 ppm
- Trace Impurities - Potassium (K):≤6.000 ppm
- Trace Impurities - Strontium (Sr):≤2.000 ppm
- Particle Count - 1.0 µm and greater:≤25 par/ml
- Reported value is the average of all samples counted for this lot number,with no individual sample value exceeding the specification.:
- NOTE: Bottle will discolor with age.:
- J.T.Baker®, Aluminum etch (80:15:3:2), CMOS:
Specification Test Results
About this item
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
UN:1760
:8
:II
:GHS05
:GHS07
:M.O.C. Category R
Customers Who Bought This Also Bought