VITLAB® Graduated Pitchers, Polypropylene, BrandTech
Catalog #: 83008-766
Supplier: Brandtech
Supplier #: 446941
Log in for availability
About this item
Made of durable, translucent polypropylene.
- Pitchers with molded graduations areavailable with or without overprint
- Nesting pitchers have screened overprint
- Blue printing provides high contrast with many liquids
Standard graduated pitchers have molded graduations with or without a blue overprint for high legibility. Nesting pitchers conserve bench and storage space and have blue screened graduations for easy, accurate measurements.
Customers Who Bought This Also Bought