Cryogenic Vial Rack, 50-Place
Catalog #: 77395-380
Supplier: Heathrow Scientific
New Product
Specifications
- Color:Blue
- For Tubes:2 - 5 ml tubes, 50 wells (12.9 mm well Ø)
Specifications
About this item
Safely store and transport cryogenic vials.
- Made from polycarbonate
- Well tabs allow one-handed opening and closing of vials that have a slot feature
- Embossed index numbers and squared, upper left corner provide easy orientation
- Drainage holes in each well aid in cleaning
- Alphanumeric identification for quick location and retrieval of tubes
- Embossed index numbers and squared, upper left corner provide easy orientation
- Ribbed grips make handling with gloves easier
The cryogenic vial rack safely stores and transports cryogenic vials.
Customers Who Bought This Also Bought