Accessories for Little Giants Washing Machines and Dryers
Supplier: Miele
New Product
Certificates
About this item
- Stainless steel stacking kit provides secure installation as a washer-dryer stack
- Stainless steel bases aid ergonomic loading and unloading of the washing machine and tumble dryer
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.