PYREX® Heavy-Duty Griffin Beakers, Graduated
Catalog #: 13912-524
Supplier: Corning
Supplier #: 1003-250
Product available on GSA Advantage®
Log in for availability
Specifications
- Catalog No:
- 13912-524
- 13912-568
- 13912-546
- 89090-528
- 89090-526
- 13912-604
- 13912-502
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
Specifications
About this item
PYREX® Heavy-Duty Griffin beakers are made of PYREX® borosilicate glass.
- Very good chemical resistance
- Thermal shock resistance
- With extra large marking spot
Heavy-duty beakers are specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. PYREX® Heavy-Duty Griffin beakers in sizes 250 ml and larger have a double graduated metric scale to indicate approximate content.
Certifications: Meet ASTM E960 specifications.
Customers Who Bought This Also Bought