ContactUsLinkComponent
VWR® Stopwatch, ¹/₁₀ sec.
VWR® Stopwatch, ¹/₁₀ sec.
Catalog #: 62387-205
Supplier:  VWR International
Small Business Enterprise
VWR* Stopwatch, 1/10 Second
VWR® Stopwatch, ¹/₁₀ sec.
Catalog #: 62387-205
Supplier:  VWR International
Small Business Enterprise
Specifications
  • Cat. no.:
    62387-205
  • Diameter:
    5.5 cm (2⁵/₃₂")
Specifications
About this item
Compact, rugged, and simple to use, with time-out and safety reset features.
  • Timing to 0.1 second, plus time-out
  • Color-coded, push-button control
  • Shock-protected, pin-lever movement
  • ABS plastic case
All push-buttons are logically color-coded to minimize timing error and ensure efficient use — green for start, red for stop, and black for return to zero position.
Instant flyback function resets and starts immediately.
The 30-second dial is graduated in tenths of a second, and a 0 to 15 minute register is color-contrasted for easy reading.
Customers Who Bought This Also Bought