Phlebotomy/Sample Collection Tray Kits, Blue/Green
Supplier: VWR International
New Product
Certificates
About this item
Keeping a clean and organized tray has never been easier. Wash the ABS tray as needed and replace the insert periodically for a fresh start.
- Multiple compartments keep a variety of supplies neatly to hand and organized
- Built in side pockets offer additional storage space
- Handle includes soft rubber grip to reduce hand and finger strain
- Each tray includes either a 13 or 16 mm Heathrow OneRack® tube rack and two disposable polystyrene inserts
- Ratcheting handle folds out of the way for easy access, even over tall sharps containers
- Includes side pockets and supply drawer
The phlebotomy tray is designed with phlebotomists in mind as it is lightweight, roomy and easy to clean. The ABS tray is washable or you can replace the insert periodically for a fresh start. Tray inserts come in two configurations and include an extra deep compartment for a sharps container.
Customers Who Bought This Also Bought