Coated Wire Centrifuge Bottle Rack
Catalog #: 77395-236
Supplier: Heathrow Scientific
New Product
Specifications
- Color:Blue
- Material:HDPE
- No. of Places:6 (2×3 array)
- Hole Size:55 - 60 mm Ø
Specifications
About this item
Chemically resistant HDPE coated wire racks.
- Holds all brands and types of conical, round, or flat bottom glass or plastic bottles
- Square wells and three deck levels help keep tubes secure
Durable for transport or sample handling. It is also chemically resistant and suitable for conical, round, or flat-bottom glass or plastic bottles.
Customers Who Bought This Also Bought