Order Entry
ContactUsLinkComponent
Coated Wire Tube Rack
Coated Wire Tube Rack
Catalog #: 77395-234
New Product
RACK TUBE COATED WIRE 16-18MM 3X5 FORMAT
Coated Wire Tube Rack
Catalog #: 77395-234
New Product

Specifications

  • Color:
    Blue
  • Material:
    HDPE
  • No. of Holes:
    15 (3×5 array)
  • For Use With:
    15 ml tubes
  • Well Shape:
    Square, 16 - 18 mm Ø

Specifications

About this item

Chemically resistant HDPE coated wire rack.

  • Holds all brands and types of conical, round, or flat bottom glass or plastic tubes
  • Square wells and three deck levels help keep tubes secure

Durable for transport or sample handling. It is also chemically resistant and suitable for conical, round, or flat bottom glass or plastic tubes.

Customers Who Bought This Also Bought