IPTG (isopropyl-β-D-thiogalactopyranoside)
Supplier: CUBE BIOTECH
New Product
Certificates
About this item
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol
Formula:C₉H₁₈O₅S
MW:238.31 g/mol
MDL Number:MFCD00063273
CAS Number:367-93-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 109.5 to 110.5 °C
:Flash Pt:
:Flash Pt:
:M.W. 238.31 g/mol
:402
Specification Test Results
- IPTG (isopropyl-β-D-thiogalactopyranoside):
Specification Test Results
Customers Who Bought This Also Bought