Nanodisc Mouse MSP1D1-His DMPC
Catalog #: 77392-794
Supplier: CUBE BIOTECH
New Product
CAS Number:
Specifications
- Size:50 µl
- Cat. no.:77392-794
Specifications
About this item
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.
- Origin of MSP protein: Mouse
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 214
- Molecular mass: 24928 Da
- Extinction coefficient (in water) ε280: 22460 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 mouse creates nanodiscs of ∼9 to 10 nm in diameter. Note that the MSP proteins in these nanodiscs are His-tagged. Keep that in mind when tagging your membrane protein of interest.
There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.
Customers Who Bought This Also Bought