Nanodisc MSP1D1 dH5-His DMPC
Catalog #: 77392-716
Supplier: CUBE BIOTECH
New Product
CAS Number:
Specifications
- Size:50 µl
- Cat. no.:77392-716
Specifications
About this item
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.
- Origin of protein: Human
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: >217
- Molecular mass: 25309 Da
- Extinction coefficient (in water) ε280: 25440 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
- Sequence (1-Letter code): MGSSHHHHHH ENLYFQ GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dH5 (His-tagged) creates nanodiscs of ∼7 to 8 nm in diameter.
This product contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.
Delivery: Shipping temperature: Dry-ice; Storage of reconstituted protein: –80 °C, avoid freeze-thaw cycles.
Customers Who Bought This Also Bought