Specifications
- Size:4×3×50 mg
- Cat. no.:77392-442
Specifications
About this item
SMALP BZs are synthesized via RAFT polymerization, achieving a better-defined polymer size and a low dispersity.
- Usage: Membrane protein solubilization and stabilization
- Full-Name: Poly(styrene-co-maleic acid-co-X%(N-benzyl)maleimide) copolymer, sodium salt in 50 mM HEPES, pH 7.5; X is be replaced with the number in the products name
- Formula Weight: 5500 g/mol
- Buffer: 50 mM HEPES
- pH: 7.5
- State: Lyophilized powder
- Recommended concentration range upon use: 1 to 5%
- Solubility: >10% in H₂O
- Color: White to slightly yellow
- Odor: Odorless
The versatility of membrane proteins and their composition mirrors the intricate nature of the cell membrane. As a result, it is a challenge to predict with absolute certainty the optimal SMALP BZ polymer for the solubilization and subsequent stabilization of your membrane protein of interest. Therefore, we offer the SMALP BZ Screening Kit that consists of our four individual SMALP BZs. This way you can conveniently screen for the best SMALP BZ for your protein before scaling up your assay. The SMALP BZ series was developed in collaboration with Prof. Bert Klumperman from Nanosene, a spinout company of Stellenbosch University in South Africa.