Ice Pan Cool Container, 4 L, Blue
Catalog #: 77395-440
Supplier: Heathrow Scientific
New Product
Specifications
- Capacity:4 L
- Color:Blue
- Description:Ice pan cool container
Specifications
About this item
Durable containers for chilling temperature-sensitive samples.
- Suitable for low temperature use.; ideal for ice, ice slurries and CO₂ pellets
- Made from polyurethane with metal inserts for stability
- Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
- Tapered design for easy stacking
- Precise pour spout and large handles
- Lid nests under bucket for convenience and to save space
- Large handles for safer transportation
Customers Who Bought This Also Bought