PVC Cryogenic Cane Sleeve, Clear
Catalog #: 77395-374
Supplier: Heathrow Scientific
New Product
Specifications
- Color:Clear
- Description:PVC cryogenic cane sleeve
Specifications
About this item
For extra security and sample safety.
- Clear plastic sleeves protect vials in aluminum canes during handling and storage
- Safe for cryogenic freezing
- More durable than cardboard sleeves
- Transparency allows sample identification without removing sleeve
The PVC cryogenic cane sleeve adds extra security and sample safety to protect vials in aluminum canes during handling and storage.
Customers Who Bought This Also Bought