Slide Staining Tray, 18-Place
Catalog #: 77395-296
Supplier: Heathrow Scientific
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Color:Black
- Description:18-place slide staining tray
- Cat. no.:77395-296
Specifications
About this item
Ideal for clinical and research laboratory staining.
- Robust design
- Durable construction with solvent resistant polypropylene blend
- Non-skid silicone feet keep tray in place
- Convenient to use
- Two ways to empty stain liquid: Pouring spouts on each corner or drain hole with plug
- Equipped with handle cut outs for easy handling
- Versatile for multiple testing formats
- Black lid included for light sensitive applications
- Humidity retention wells for specialized staining needs
Tray holds 18 standard microscope slides (76×26 mm) securely in place for staining, rinsing and drying applications.
Customers Who Bought This Also Bought