Specifications
- Cat. no.:77392-708
- Contents:MSP2N2-His lyophilized protein 2 mg; Palmitoyl-oleoyl-phosphatidylcholine (POPC) 9 mg; Sodium cholate 2×20 mg
Specifications
About this item
This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP2N2 (His-tagged) and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.
- Origin of protein: Human
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: >217
- Molecular mass: 25309 Da
- Extinction coefficient (in water) ε280: 25440 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Sequence (1-Letter Code): MGSSHHHHHH ENLYFQ GLKLLDNWDSVTSTFSKLREQLG PVTQEFWDNLE KETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFK VSFLSALEEY TKKLNTQ
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 (His-tagged) creates nanodiscs of ∼17 nm in diameter.
With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs. Just use the corresponding protocol for each purpose (see below). The MSP protein contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein.