Nanodisc Assembly Kit MSP1D1 dH5 POPC
Catalog #: 77392-700
Supplier: CUBE BIOTECH
New Product
CAS Number:
Specifications
- Cat. no.:77392-700
- Contents:MSP1D1ΔH5 lyophilized protein 2 mg; Palmitoyl-oleoyl-phosphatidylcholine (POPC) 3.12 mg; Sodium cholate 2×20 mg
Specifications
About this item
This product is a MSP nanodisc assembly kit. It contains a vial of lyophilized membrane scaffold protein MSP1D1 dh5 and a vial of DMPC or POPC phopshospholipid to assemble a MSP nanodisc.
- Origin of MSP protein: Human
- Purity: >90% (determined by SDS-PAGE)
- Number of amino acids: 392
- Molecular mass: 45670 Da
- Extinction coefficient (in water) ε280: 44920 M⁻¹ cm⁻¹
- Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
- Sequence (human origin): GHHHHHHH DYDIPTT ENLYFQG STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 dh5 creates nanodiscs of ∼7 to 8 nm in diameter.
With this assembly kit both cellular as well as cell-free expressed proteins can be stabilized in MSP nanodiscs.
Delivery: Shipping temperature: Room temperature; storage of lyophilized copolymer: −20 °C for several years; storage of reconstituted protein: 2 - 8 °C for several day.
Customers Who Bought This Also Bought