Specifications
- Size:50 µl
- Cat. no.:77392-820
Specifications
About this item
Membrane scaffold protein; rat origin.
- Usage: Specific binding and purification of 6× His-tagged proteins
- Specificity: Affinity to His-tagged proteins
- Binding capacity: >70 mg/ml
- Bead ligand: Ni-IDA
- Bead size: 30 μm
- Filling quantity: Delivered as a 25% suspension
- pH stability: 2 to 14
- Chelator stability: Stable in buffer containing 10 mM DTT and 1 mM EDTA
- Other stabilities: 100% methanol, 100% ethanol, 8 M urea, 6 M guanidinium hydrochloride, 30% (v/v) acetonitrile
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 rat creates nanodiscs of ∼9 to 10 nm in diameter.
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein. There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.
Note that the MSP proteins in these nanodiscs are His-tagged. Keep that in mind when tagging your membrane protein of interest.