Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Methyl cyanide,Ethanoic acid nitrile,Cyanomethane
Formula:H₃CCN
MW:41.05 g/mol
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
MDL Number:MFCD00001878
CAS Number:75-05-8
ADR:3,II
Merck Index:13,00071
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Boiling Pt:
:Boiling Pt: 81.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -45.7 °C|Melting Pt: –45.7 °C
:Flash Pt:
:Flash Pt: 2 °C (closed cup)
:M.W. 41.05 g/mol
:Acetonitrile
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
REACH:01-2119471307-38
:3
:II
:2 °C
:GHS02
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):99.9% min
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):300ppt max
- Form:Clear liquid
- Gradient at 210 nm:0.002AU max
- Gradient at 254 nm:0.0003AU max
- Identity (IR-spectrum):Conforms
- Residue after evaporation:1ppm max
- Titrable acid:8.0µeq/g max
- Titrable base:0.16µeq/g max
- UV Abs. at 189 nm:1.00AU max
- UV Abs. at 195 nm:0.15AU max
- UV Abs. at 210 nm:0.03AU max
- UV Abs. at 230 nm:0.005AU max
- UV Abs. at 250 nm:0.005AU max
- UV Abs. at 270 nm:0.005AU max
- UV Cut-off:189nm max
- Water (H2O):0.02% max
- Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®:
Specification Test Results
Related Information
Customers Who Bought This Also Bought