Nanodisc Rat MSP1E3D1-His POPC
Catalog #: 77392-828
Supplier: CUBE BIOTECH
New Product
CAS Number:
Specifications
- Size:50 µl
- Cat. no.:77392-828
Specifications
About this item
Membrane scaffold protein. Rat origin.
- Origin of the MSP protein: Rat
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 267
- Molecular Mass: 31572.76 Da
- Extinction coefficient (in water) ε280: 36440 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (rat) creates nanodiscs of ∼12 to 14 nm in diameter. This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein. There are multiple phospholipids to choose from that can make up the body of the nanodisc. Which is best, depends on your protein.
Delivery: Shipping Temperature: Dry-Ice. Storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.
Customers Who Bought This Also Bought