Nanodisc MSP2N2 POPC
Catalog #: 77392-754
Supplier: CUBE BIOTECH
New Product
CAS Number:
Specifications
- Size:50 µl
- Cat. no.:77392-754
Specifications
About this item
This product is a pre-assembled nanodisc. Its intended purpose is to stabilize a cell-free expressed membrane protein.
- Origin of the MSP protein: Human
- Purity: >90% (Determined by SDS-PAGE)
- Number of amino acids: 371
- Molecular Mass: 43.05 kDa
- Extinction coefficient (in water) ε280: 36900 M⁻¹cm⁻¹
- Buffer: 20 mM TRIS pH 7.4, 100 mM NaCl, 0.5 mM EDTA
- Sequence (1 Letter Code): G STFSKLREQLG PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ GT PVTQEFWDNLEKETEGLRQEMS KDLEEVKAKVQ PYLDDFQKKWQEEMELYRQKVE PLRAELQEGARQKLHELQEKLS PLGEEMRDRARAHVDALRTHLA PYSDELRQRLAARLEALKENGG ARLAEYHAKATEHLSTLSEKAK PALEDLRQGLL PVLESFKVSFLSALEEYTKKLNTQ
- Affinity tag: none
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP2N2 creates nanodiscs of ∼17 nm in diameter.
Delivery: Shipping Temperature: Dry-Ice. Storage of reconstituted protein: −80 °C, avoid freeze-thaw cycles.
Customers Who Bought This Also Bought