About this item
Large volume magnetic stirrers are designed for large capacity applications
- 230 V model
Powerful rare earth magnets automatically pull stir bars into position and stay coupled under the most demanding stirring conditions. The rugged ABS/polycarbonate housing and heavy-duty motor offer exceptional magnetic force and provide extended reliability. Stirrers feature non-skid feet to prevent the unit from moving during operation. The MegaMag Genie™ stirrers feature simple analog controls that allow user to easily change the desired speed. The digital MegaMag Genie™ stirrers feature true speed readout with feedback control that is adjustable in 10 rpm increments. Digital unit also includes an adjustable timer. The analog and digital MegaMag Genie™ stirrers can stir up to 38 L (10 gal.)
Specifications
- Catalog No:
- 89037-474
- 89037-470
- Weight:
- 3 kg (6.6 lbs.)
- 3 kg (6.6 lbs.)
- Overall dimensions:
- 13H×30.5W×31D cm (51/8×12×127/32")
- 13H×30.5W×31D cm (51/8×12×127/32")