Precision Balances, Standard MA
Supplier: Mettler Toledo
New Product
Certificates
About this item
METTLER TOLEDO Standard MA Precision Balances have a large weighing pan, robust design and highly accurate weigh cell with internal adjustment.
- Statistical data analysis
- Front level indicator
- Metal base, PBT top housing
- Overload protection
- Two interfaces: USB-A, RS232
- Bluetooth option
The hybrid LCD touchscreen increases efficiency in the workflow with intuitive onscreen guidance and user management. Easily navigate 10 built-in applications and export weighing data. Built with robust construction to defend against external influences and provide high quality service for years to come. Easily export weighing data to a computer, printer, or USB storage device.
Specifications
- Catalog No:
- 77323-256
- CA77323-248
- CA77323-244
- CA77323-256
- CA77323-252
- 77323-244
- 77323-248
- 77323-252
- Communication interface:
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- USB-A, RS232
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz