Precision Balances, Legal For Trade, Advanced MX
Supplier: Mettler Toledo
New Product
Certificates
About this item
METTLER TOLEDO Advanced MX Legal for Trade Precision Balances feature a precise weigh cell and automatic internal adjustment. The SmartPan stabilizes in any weighing environment, ensuring consistently accurate results.
- Statistical data analysis
- Leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
Exceptional user management within the 7" touchscreen allows easy access to 9 built-in weighing applications, assigns user access rights and provides automatic data logging. These balances are designed with full metal housing and IP54 protection to defend against environmental influences and rough handling. The power-saving mode can be enabled when the balance is not in use to save costs and reduce energy usage.
Certifications: OIML/NTEP Approved.
Specifications
- Catalog No:
- 77323-066
- 77323-094
- 77323-062
- 77323-078
- 77323-070
- 77323-098
- 77323-082
- 77323-074
- 77323-102
- 77323-086
- 77323-106
- 77323-090
- 77323-110
- Communication interface:
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz