Advanced MX Precision Balances
Supplier: Mettler Toledo
New Product
Certificates
About this item
METTLER TOLEDO Advanced MX Precision Balances feature a large weighing pan, robust design, and precise weigh cell with FACT automatic internal adjustment.
- SmartPan
- Statistical data analysis
- User management; Leveling assistant
- StatusLight
- 3 interfaces: USB-A, USB-C, Ethernet
- Bluetooth option
- EasyDirect software
- Full metal housing with overload protection
The 7" intuitive touchscreen with user management provides automatic data logging, statistical analysis and easy access to 9 built-in weighing applications. Transfer data to a printer or computer for error-free documentation. Built with IP 54 protection and robust construction, these balances will deliver high quality performance for years to come. Enable the power-saving mode when balance is not in use to saves costs and reduces energy consumption.
Specifications
- Catalog No:
- CA77323-112
- CA77323-116
- 77323-112
- 77323-116
- 77323-120
- CA77323-120
- 77323-124
- CA77323-124
- Communication interface:
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- USB-A, USB-C, Ethernet
- Voltage:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz