Hemostats for Soldering, Aven Tools
Supplier: Aven Tools
Certificates
Certificates
About this item
Versatile tools for use as clamps, heat sinks, third hands, and more while soldering
With three-position locking device. Stainless steel construction.
Customers Who Bought This Also Bought
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
$200.31 to $2,030.49
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
$768.73 to $3,075.66