RNA Viral Infection RT-PCR Kit
Catalog #: 470355-108
Supplier: GENOSENSOR CORPORATION
New Product
Specifications
- Description:RNA viral infection RT-PCR kit
- No. of Lab Groups:6 groups of 4
- Cat. no.:470355-108
Specifications
About this item
The RNA virial infection RT-PCR kit is designed to introduce students to the RNA viral world and RT-PCR – an RNA amplification technology.
- Perform an RNA amplification via RT-PCR technique
- Verify presence of RT-PCR amplified RNA by gel electrophoresis
- Fast and simple protocol for easy integration into biotechnology lessons
In this lab, students play the role of the diagnostician in the following scenario. Patients arrive at the emergency room with fever and cough. An RNA viral infection is suspected and must be confirmed or ruled-out from patient samples. A short RNA is used to emulate the virus; no actual viral particles are used.
Customers Who Bought This Also Bought