Four-Edged Glass Scoring Knife, Wale Apparatus
Catalog #: 32861-011
Supplier: Wale Apparatus
Specifications
- Description:Glass Scoring Knife
- Cat. no.:32861-011
- Supplier No.:90-6050
Specifications
About this item
The four cutting edges of this knife ensures a greater ease of use and longer life and it is ideally designed for cutting glass and quartz tubing and rods.
- Retractable and replaceable blade
Unique square collet holds high-quality tungsten-carbide steel blade. User can expose as much of blade as desired and easily replace a worn blade.
Ordering information: Knife comes complete with four-edged blade. Replacement blades available.
Customers Who Bought This Also Bought