Specifications
- Description:3Doodler Pro+ 3D printing pen set
- Cat. no.:470354-042
Specifications
About this item
A professional-grade 3D printing solution.
- Robust dual-drive system built to run longer and harder than any other 3D pen
- Versatile enough to accept ABS, PLA, Metal, FLEXY, and Nylon filaments
- Plug-in power supply and temperature selection options ensure the Pro+ pen operates at the optimal temperature
The 3Doodler Pro+ combines sleek, ergonomic styling with advanced technology for superior performance to elevate design and bring ideas to life. Explore 3D design and prototypes in a fraction of the time and share ideas using a range of materials from ABS to metal and nylon — the possibilities are endless.
The PRO+ rests comfortably in the hand and features a magnetic maintenance panel that enables easy access the pen’s interior. The digitally adjustable speed and temperature settings offer unparalleled design control. Create professional-grade Doodles with 6 packs of refills, including ABS, PLA, Metal, Flexy, and Nylon, equivalent to 125 feet of plastic and over 1,500 feet of extrusion.
There is also a Nozzle Set included which enables you to swap out between six different nozzle tips to customize your outcome. It includes a ribbon tip, square tip, triangle tip, a super fine 0.5 mm nozzle, and thicker 1 mm and 1.5 mm nozzles.