About this item
Sterile coveralls are made from a comfortable fabric that protects against various liquids and particulates. Each coverall is individually packaged and sterilized to a 10⁻⁶ sterility assurance level (SAL).
- Attached hood and boots eliminates the need for donning and storing separate hood and boot components
- Attached thumb loops help prevent sleeves from riding up
- Ankle ties provide customized fit for boots
- Tunnelized elastic wrists, ankles, and back; serged seams
- Sterile-Don™ packaging protects against contamination during donning
- GammaGuard CE® helps keep wearer cooler compared to competitive fabrics
- Meets high manufacturing standards; made in an ISO-certified facility
Coveralls are designed with tunnelized elastic that contains particle shed. Serged seams eliminate raw edges that can create and release additional particulate contamination. Work confidently with an attached hood and boots that provide more complete body protection. Elastic wrists and ankles help prevent particulates from entering or exiting the suit, while a storm flap over the front zipper closure adds an extra layer of protection. Convenient thumb loops are included to keep sleeves from riding up while ankle ties secure the attached boots.