UltraSOLV® Cleanroom Sponge
Catalog #: 77394-794
Supplier: Foamtec International
New Product
Specifications
- W×L:2.62×4.38"
- Color:White
- Description:UltraSOLV® sponge
Specifications
About this item
Using Foamtec's patented cleaning process and foam technology, UltraSOLV® sponges are ideal for cleaning critical tools and assemblies. The sponge can be used with UltraSOLV® ScrubPADs on wet cleans.
- Constructed from a naturally reticulated, hydrophilic, polyurethane foam
- Unique abrasion resistance allows for cleaning rough, coarse, and bead blasted sufaces
- Static dissipative and tribocharging properties
- For use with static sensitive equipment
- Class 10/ISO 4 compatible
Fiber-free hydrophilic polyurethane foam construction combined with low levels of particle generation, ionics, and organics make these sponges ideal for for wet cleaning of process equipment in the semiconductor, disk drive, and flat-panel display industries.
Packaging: 100 units per case.
Customers Who Bought This Also Bought