ContactUsLinkComponent
ADSC Basal Medium
ADSC Basal Medium
Catalog #: CA10064-096
CAS Number:  
Media, ADSC - Apidose-Derived Stem Cells Basal Medium, 500 ml
ADSC Basal Medium
Catalog #: CA10064-096
CAS Number:  
Specifications
  • Cat. no.:
    CA10064-096
Specifications
About this item
Stem cells possess the unique ability to proliferate as an undifferentiated cell type, then, when properly stimulated, differentiate into multiple lineages of cells.
Lonza’s ADSC human Adipose Derived Stem Cell Growth BulletKit® Medium is a serum containing medium designed to proliferate human adipose derived mesenchymal stem cells (both normal and diseased) in an undifferentiated state.
The serum used in Lonza’s ADSC human Adipose Derived Stem Cell Growth BulletKit® Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).
The ADSC human Adipose Derived Stem Cell Growth BulletKit® Medium is offered as a BulletKit® Medium (catalog no. PT-4505) which includes both the basal media and the necessary supplements for proliferation of human adipose derived mesenchymal stem cells. The basal medium (ADSC Basal Medium; catalog no. PT-3273) and the necessary supplements (ADSC-GM SQ Kit; catalog no. PT-44503) can also be purchased individually.
Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed.
Delivery: Content: ADSC Basal Medium (1 bottle).
Customers Who Bought This Also Bought