Specifications
- Description:MSCBM® Mesenchymal stem cell basal medium
- Cat. no.:CA10064-094
- Supplier No.:PT-3238
Specifications
About this item
MSCBM Basal Medium is the basal medium component of the MSCGM human Mesenchymal Stem Cell Growth BulletKit® (catalog no. PT-3001),a serum containing medium designed to proliferate human bone marrow derived mesenchymal cells in an undifferentiated state.
The serum used in Lonza’s MSCGM®human Mesenchymal Stem Cell Growth Medium BulletKit® Medium is specifically screened to ensure that the serum neither promotes spontaneous differentiation of cells nor inhibits cell differentiation to osteogenic, chondrogenic, or adipogenic lineages (when properly stimulated with an appropriate differentiation media).
Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed.