BulletKit® NPDM® Neural Progenitor Differentiation Medium
Catalog #: CA10063-894
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Cat. no.:CA10063-894
Specifications
About this item
Poietics® Neural Progenitor Cell System contains Norman Human Neural Progenitor Cells (NHNP) and defined medium optimized for their growth and in vitro differentiation.
This system may be used for experimental applications including drug development, neurotoxicity, neurogenesis, electrophysiology, neurotransmitter disorders, and central nervous system function.
The available defined neural progenitor media includes a growth medium BulletKit® (CC-3209) as well as a differentiaion medium BulletKit® (CC-3229).
Poietics® Cells, Medium and Reagents are quality tested together and guaranteed to give optimum performance as a complete cell system.
Customers Who Bought This Also Bought