ContactUsLinkComponent
LGM-3® Lymphocyte Growth Medium-3
LGM-3® Lymphocyte Growth Medium-3
Catalog #: CA10064-356
CAS Number:  
Clonetics* LGM-3 Growth Medium 500 ml
LGM-3® Lymphocyte Growth Medium-3
Catalog #: CA10064-356
CAS Number:  
Specifications
  • Description:
    LGM-3® Lymphocyte growth medium-3
  • Cat. no.:
    CA10064-356
  • Supplier No.:
    CC-3211
Specifications
About this item
LGM-3® Lymphocyte Growth Medium is a chemically defined, serum-free, xeno-free media developed to support (with the addition of appropriate cytokines) the growth and support of human lymphocytes and dendritic cells.
LGM-3® Lymphocyte Growth Medium does not contain any exogenous growth factors, artificial stimulators of cellular proliferation, or undefined supplements.
LGM-3® Lymphocyte Growth Medium is devoid of any protein kinase C stimulators and is suitable for the investigation of second messenger systems in the activation of human lymphocytes. LGM-3® Lymphocyte Growth Medium's formulation is complete and contains human albumin, recombinant human insulin, and pasteurized human transferrin. LGM-3® Lymphocyte Growth Medium may be further supplemented with additional cytokines and growth factors depending on applications.
Lonza guarantees the performance of Clonetics®/Poietics® cells only if appropriate Clonetics®/Poietics® media and reagents are used exclusively and the recommended storage and use protocols are followed.
Delivery: Content: 1 × LGM-3® Lymphocyte Growth Medium-3, 500 ml.
Customers Who Bought This Also Bought